Anti-beta Amyloid Mouse Monoclonal Antibody [clone: 16E9]

Supplier: Novus Biologicals
NBP2-23544
103404-918EA 383 USD
103404-918
Anti-beta Amyloid Mouse Monoclonal Antibody [clone: 16E9]
Antibodies
The beta Amyloid Antibody (16E9) - BSA Free from Novus Biologicals is a mouse monoclonal antibody to beta Amyloid. This antibody reacts with human. The beta Amyloid Antibody (16E9) - BSA Free has been validated for the following applications: ELISA.

Type: Primary
Antigen: beta Amyloid
Clonality: Monoclonal
Clone: 16E9
Conjugation: Unconjugated
Epitope: Epitope specificity differs from that of Clone 03 and Clone 07
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR