Anti-beta Amyloid Mouse Monoclonal Antibody (DyLight 488) [clone: 12C3]

Supplier: Novus Biologicals
NBP1-05103G
103402-708EA 399 USD
103402-708
Anti-beta Amyloid Mouse Monoclonal Antibody (DyLight 488) [clone: 12C3]
The beta Amyloid Antibody (12C3) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to beta Amyloid. This antibody reacts with non-species specific. The beta Amyloid Antibody (12C3) [DyLight 488] has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: beta Amyloid
Clonality: Monoclonal
Clone: 12C3
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR