Anti-beta Adducin Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00000119-B01P
103321-422EA 399 USD
103321-422
Anti-beta Adducin Mouse Polyclonal Antibody
Antibodies
The beta Adducin Antibody from Novus Biologicals is a mouse polyclonal antibody to beta Adducin. This antibody reacts with human. The beta Adducin Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: beta Adducin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR