Anti-beta 2-Microglobulin Mouse Monoclonal Antibody [clone: 3G5H8]

Supplier: Novus Biologicals
NBP2-37287
103402-640EA 635.51 USD
103402-640
Anti-beta 2-Microglobulin Mouse Monoclonal Antibody [clone: 3G5H8]
Antibodies
The beta 2-Microglobulin Antibody (3G5H8) from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human. The beta 2-Microglobulin Antibody (3G5H8) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF-ready.

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: 3G5H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR