Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 488) [clone: TLD-3H12B]

Supplier: Novus Biologicals
NB600-1383G
103404-896EA 607.2 USD
103404-896
Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 488) [clone: TLD-3H12B]
Antibodies
The beta 2-Microglobulin Antibody (TLD-3H12B) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with rat. The beta 2-Microglobulin Antibody (TLD-3H12B) [DyLight 488] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: TLD-3H12B
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR