Anti-beta 2-Microglobulin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15566
102706-234EA 635.51 USD
102706-234
Anti-beta 2-Microglobulin Rabbit Polyclonal Antibody
Antibodies
The beta 2-Microglobulin Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta 2-Microglobulin. This antibody reacts with human, mouse, rat. The beta 2-Microglobulin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR