Anti-beta 2-Microglobulin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
22110002-0.1MG
103317-716EA 349 USD
103317-716
Anti-beta 2-Microglobulin Rabbit Polyclonal Antibody
Antibodies
The beta 2-Microglobulin Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta 2-Microglobulin. This antibody reacts with human. The beta 2-Microglobulin Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary Antibodies
Antigen: beta 2-Microglobulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR