Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 488) [clone: B2M/961]

Supplier: Novus Biologicals
NBP2-47704G
103404-714EA 638.77 USD
103404-714
Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 488) [clone: B2M/961]
Antibodies
The beta 2-Microglobulin Antibody (B2M / 961) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human, primate. The beta 2-Microglobulin Antibody (B2M / 961) [DyLight 488] has been validated for the following applications: Flow Cytometry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: B2M/961
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR