|
![]() |
ORDER
SPECIFICATIONS
Antigen Name | beta 2-Microglobulin |
Reactivity | Primate, Human |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | B2M/961 |
Host | Mouse |
Isotype | IgG2b Kappa |
Flow Cytometry | Yes |
Antigen Synonyms | beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin |
Gene ID | 567 |
Immunogen | Full length recombinant human B2M protein |
Purification | Protein A or G purified |
Storage Buffer | PBS |
Storage Temperature | Store at 4C in the dark. |