Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 550) [clone: SPM617]

Supplier: Novus Biologicals
NBP2-47703R
103402-680EA 638.77 USD
103402-680
Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 550) [clone: SPM617]
Antibodies
The beta 2-Microglobulin Antibody (SPM617) [DyLight 550] from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human, primate. The beta 2-Microglobulin Antibody (SPM617) [DyLight 550] has been validated for the following applications: Flow Cytometry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: SPM617
Conjugation: DyLight 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR