Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 650) [clone: B2M/1118]

Supplier: Novus Biologicals
NBP2-47702C
103404-248EA 638.77 USD
103404-248
Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 650) [clone: B2M/1118]
Antibodies
The beta 2-Microglobulin Antibody (B2M / 1118) [DyLight 650] from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human, primate. The beta 2-Microglobulin Antibody (B2M / 1118) [DyLight 650] has been validated for the following applications: Flow Cytometry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: B2M/1118
Conjugation: DyLight 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR