Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 755) [clone: SPM374]

Supplier: Novus Biologicals
NBP2-47701IR
103402-664EA 429 USD
103402-664
Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 755) [clone: SPM374]
Antibodies
The beta 2-Microglobulin Antibody (SPM374) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human, primate (negative). The beta 2-Microglobulin Antibody (SPM374) [DyLight 755] has been validated for the following applications: Flow Cytometry.

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: SPM374
Conjugation: DyLight 755
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Primate (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR