Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 550) [clone: 246-E9.E7 (same as HLA.ABC.m2)]

Supplier: Novus Biologicals
NBP2-47700R
103404-670EA 638.77 USD
103404-670
Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 550) [clone: 246-E9.E7 (same as HLA.ABC.m2)]
Antibodies
The beta 2-Microglobulin Antibody (246-E9.E7 (same as HLA.ABC.m2)) [DyLight 550] from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human, primate (negative). The beta 2-Microglobulin Antibody (246-E9.E7 (same as HLA.ABC.m2)) [DyLight 550] has been validated for the following applications: Flow Cytometry.

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: 246-E9.E7 (same as HLA.ABC.m2)
Conjugation: DyLight 550
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Primate (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR