Anti-beta 2-Microglobulin Mouse Monoclonal Antibody [clone: B2M/1118]

Supplier: Novus Biologicals
NBP2-48327
103404-704EA 515.69 USD
103404-704
Anti-beta 2-Microglobulin Mouse Monoclonal Antibody [clone: B2M/1118]
Antibodies
The beta 2-Microglobulin Antibody (B2M / 1118) - IHC-Prediluted from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human, primate. The beta 2-Microglobulin Antibody (B2M / 1118) - IHC-Prediluted has been validated for the following applications: Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: B2M/1118
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR