Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Basal cell cytokeratin Mouse Monoclonal Antibody (DyLight 488) [clone: RCK103]

Supplier: Novus Biologicals
The Basal cell cytokeratin Antibody (RCK103) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to Basal cell cytokeratin. This antibody reacts with human, rat, porcine, avian, canine, chicken, guinea pig, hamster, rabbit, zebrafish. The Basal cell cytokeratin Antibody (RCK103) [DyLight 488] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Basal cell cytokeratin
Clonality: Monoclonal
Clone: RCK103
Conjugation: DyLight 488
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Porcine, Avian, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-97729G 103405-330 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Name Basal cell cytokeratin
Reactivity Human, Hamster, Guinea Pig, Chicken, Canine, Rabbit, Porcine, Rat, Avian, Zebrafish
Conjugation DyLight® 488
Clonality Monoclonal
Antibody Type Primary
Clone RCK103
Host Mouse
Isotype IgG1
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Immunogen RCK103 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a mouse immunized with a mix of cell preparations containing human cytokeratins.
Purification Protein A or G purified
Storage Buffer 50mM Sodium Borate
Storage Temperature Store at 4C in the dark.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty