|
![]() |
ORDER
SPECIFICATIONS
Antigen Name | Basal cell cytokeratin |
Reactivity | Human, Avian, Rat, Zebrafish, Rabbit, Porcine, Hamster, Guinea Pig, Canine, Chicken |
Conjugation | PerCP (Peridinin-Chlorophyll Protein Complex) |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | RCK103 |
Host | Mouse |
Isotype | IgG1 |
Western Blot | Yes |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Flow Cytometry | Yes |
Immunogen | RCK103 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a mouse immunized with a mix of cell preparations containing human cytokeratins. |
Purification | Protein A or G purified |
Storage Buffer | PBS |
Storage Temperature | Store at 4C in the dark. |