Anti-Basal cell cytokeratin Mouse Monoclonal Antibody (DyLight 350) [clone: RCK103]

Supplier: Novus Biologicals
NBP1-97729UV
103405-324EA 777.05 USD
103405-324
Anti-Basal cell cytokeratin Mouse Monoclonal Antibody (DyLight 350) [clone: RCK103]
Antibodies
The Basal cell cytokeratin Antibody (RCK103) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to Basal cell cytokeratin. This antibody reacts with human, rat, porcine, avian, canine, chicken, guinea pig, hamster, rabbit, zebrafish. The Basal cell cytokeratin Antibody (RCK103) [DyLight 350] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Basal cell cytokeratin
Clonality: Monoclonal
Clone: RCK103
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Porcine, Avian, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR