Anti-Baculovirus Envelope gp64 protein Mouse Monoclonal Antibody (DyLight 680) [clone: AcV1]

Supplier: Novus Biologicals
NBP2-00208FR
103402-318EA 564.74 USD
103402-318
Anti-Baculovirus Envelope gp64 protein Mouse Monoclonal Antibody (DyLight 680) [clone: AcV1]
Antibodies
The Baculovirus Envelope gp64 protein Antibody (AcV1) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to Baculovirus Envelope gp64 protein. This antibody reacts with virus. The Baculovirus Envelope gp64 protein Antibody (AcV1) [DyLight 680] has been validated for the following applications: Flow Cytometry.

Type: Primary
Antigen: Baculovirus Envelope gp64 protein
Clonality: Monoclonal
Clone: AcV1
Conjugation: DyLight 680
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR