Anti-Axotrophin Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB100-93417
103405-908EA 564.74 USD
103405-908
Anti-Axotrophin Goat Polyclonal Antibody
Antibodies
The Axotrophin Antibody from Novus Biologicals is a goat polyclonal antibody to Axotrophin. This antibody reacts with human, mouse, rat. The Axotrophin Antibody has been validated for the following applications: Peptide ELISA.

Peptide

Type: Primary
Antigen: Axotrophin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-NFNQESEGRNT
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR