Anti-Avian Influenza Hemagglutinin 3 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-41065
103405-904EA 564.74 USD
103405-904
Anti-Avian Influenza Hemagglutinin 3 Goat Polyclonal Antibody
Antibodies
The Avian Influenza Hemagglutinin 3 Antibody from Novus Biologicals is a goat polyclonal antibody to Avian Influenza Hemagglutinin 3. This antibody reacts with virus. The Avian Influenza Hemagglutinin 3 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Avian Influenza Hemagglutinin 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR