Anti-Avian Influenza Hemagglutinin 1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-41069
103405-900EA 564.74 USD
103405-900
Anti-Avian Influenza Hemagglutinin 1 Goat Polyclonal Antibody
Antibodies
The Avian Influenza Hemagglutinin 1 Antibody from Novus Biologicals is a goat polyclonal antibody to Avian Influenza Hemagglutinin 1. This antibody reacts with virus. The Avian Influenza Hemagglutinin 1 Antibody has been validated for the following applications: ELISA.

Type: Primary
Antigen: Avian Influenza Hemagglutinin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR