Anti-Avian Flu Nucleoprotein Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56611SS
103360-814EA 208 USD
103360-814
Anti-Avian Flu Nucleoprotein Rabbit Polyclonal Antibody
Antibodies
The Avian Flu Nucleoprotein Antibody from Novus Biologicals is a rabbit polyclonal antibody to Avian Flu Nucleoprotein. This antibody reacts with avian, chicken. The Avian Flu Nucleoprotein Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Avian Flu Nucleoprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Avian, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR