Anti-Aurora C Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00006795-B01P
103310-952EA 798.46 USD
103310-952
Anti-Aurora C Mouse Polyclonal Antibody
Antibodies
The Aurora C Antibody from Novus Biologicals is a mouse polyclonal antibody to Aurora C. This antibody reacts with human. The Aurora C Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Aurora C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR