Anti-Aurora B Mouse Monoclonal Antibody [clone: 6G8]

H00009212-M04
103336-420EA 553.85 USD
103336-420
Anti-Aurora B Mouse Monoclonal Antibody [clone: 6G8]
The Aurora B Antibody (6G8) from Novus Biologicals is a mouse monoclonal antibody to Aurora B. This antibody reacts with human. The Aurora B Antibody (6G8) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Aurora B
Clonality: Monoclonal
Clone: 6G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR