|
![]() |
ORDER
SPECIFICATIONS
Antigen Name | Aurora A/B/C |
Reactivity | Porcine, Chicken, Rat, Bovine, Equine, Human, Mouse |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | 4A7 |
Host | Mouse |
Isotype | IgG1 |
Western Blot | Yes |
ImmunoFluorescence | Yes |
Concentration | 1 mg/ml |
Immunogen | Recombinant full length human Aurora C. [UniProt# Q9UQB9] |
Purification | Protein G purified |
Storage Buffer | PBS |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |