Anti-Aurora A/B Mouse Monoclonal Antibody [clone: 5A12]

Supplier: Novus Biologicals
NBP2-50049SS
103401-900EA 208 USD
103401-900
Anti-Aurora A/B Mouse Monoclonal Antibody [clone: 5A12]
Antibodies
The Aurora A / B Antibody (5A12) from Novus Biologicals is a mouse monoclonal antibody to Aurora A / B. This antibody reacts with human, mouse, rat, porcine, bovine, chicken, equine. The Aurora A / B Antibody (5A12) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Aurora A/B
Clonality: Monoclonal
Clone: 5A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR