Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Aurora A Mouse Monoclonal Antibody [clone: 1F8]

Supplier: Novus Biologicals
NBP2-22118SS
The Aurora A Antibody (1F8) from Novus Biologicals is a mouse monoclonal antibody to Aurora A. This antibody reacts with human, rat, primate. The Aurora A Antibody (1F8) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Aurora A
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Primate




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.025 ml NBP2-22118SS 103401-890 Each (25µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Name Aurora A
Reactivity Human, Rat, Primate
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 1F8
Host Mouse
Isotype IgG1
Western Blot Yes
ELISA Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Antigen Synonyms serine/threonine kinase 6, STK15serine/threonine kinase 15, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, aurora kinase AhARK1, AIKAurA, breast-tumor-amplified kinase, ARK1EC 2.7.11.1, serine/threonine-protein kinase 6, IPL1-related kinase, Aurora/IPL1-related kinase 1, STK7, STK6MGC34538, BTAKAURORA2, ARK-1, EC 2.7.11, Breast tumor-amplified kinase, aurora/IPL1-like kinase, Aurora-related kinase 1, AURA, Serine/threonine-protein kinase aurora-A
Gene ID 6790
Concentration 1.0 mg/ml
Immunogen Purified recombinant fragment of human Aurora A expressed in E. coli. [UniProt# O14965]
Purification Ammonium sulfate precipitation
Storage Buffer PBS
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty