Anti-Aurora A Mouse Monoclonal Antibody [clone: 1A14]

Supplier: Novus Biologicals
NBP2-50041SS
103401-886EA 121.54 USD
103401-886
Anti-Aurora A Mouse Monoclonal Antibody [clone: 1A14]
Antibodies
The Aurora A Antibody (1A14) from Novus Biologicals is a mouse monoclonal antibody to Aurora A. This antibody reacts with human. The Aurora A Antibody (1A14) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Aurora A
Clonality: Monoclonal
Clone: 1A14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR