Anti-Aubergine/Sting Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-21661
102713-344EA 777.05 USD
102713-344
Anti-Aubergine/Sting Rabbit Polyclonal Antibody
Antibodies
The Aubergine / Sting Antibody from Novus Biologicals is a rabbit polyclonal antibody to Aubergine / Sting. This antibody reacts with drosophila. The Aubergine / Sting Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Aubergine/Sting
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR