Anti-AU1 Epitope Tag Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB600-452-1ML
103405-896EA 979 USD
103405-896
Anti-AU1 Epitope Tag Goat Polyclonal Antibody
The AU1 Epitope Tag Antibody from Novus Biologicals is a goat polyclonal antibody to AU1 Epitope Tag. This antibody reacts with e. coli, non-species specific. The AU1 Epitope Tag Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: AU1 Epitope Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: E. coli, Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR