|
![]() |
ORDER
SPECIFICATIONS
Antigen Name | ATPase Na+/K+ beta 2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | sodium/potassium-transporting ATPase subunit beta-2, adhesion molecule on glia, AMOGsodium/potassium-dependent ATPase beta-2 subunit, Na, beta 2 polypeptide, K-ATPase beta-2 polypeptide, ATPase, sodium/potassium-transporting ATPase beta-2 chain, Sodium/potassium-dependent ATPase subunit beta-2, Na+/K+ transporting |
Gene ID | 482 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TVSDHTPKYQDRLATPGLMIRPKTENLDVIVNVSDTESWDQHVQKLNKFLEPYNDSIQAQKNDVCRPGRYYEQPDNGVLNYPKRACQFNRTQLGNCSGIGDST |
Purification | Immunogen affinity purified |
Storage Buffer | PBS (pH 7.2) and 40% Glycerol |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |