Anti-ATPase Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-52179
103405-894EA 564.74 USD
103405-894
Anti-ATPase Goat Polyclonal Antibody
Antibodies
The ATPase Antibody from Novus Biologicals is a goat polyclonal antibody to ATPase. This antibody reacts with human, mouse, rat, canine, rabbit. The ATPase Antibody has been validated for the following applications: Peptide ELISA.

Peptide

Type: Primary
Antigen: ATPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-SQTTHDLSKFDR
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Canine, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR