Anti-Ataxin-2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00006311-A01
103309-808EA 299 USD
103309-808
Anti-Ataxin-2 Mouse Polyclonal Antibody
Antibodies
The Ataxin-2 Antibody from Novus Biologicals is a mouse polyclonal antibody to Ataxin-2. This antibody reacts with human. The Ataxin-2 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Ataxin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR