Anti-Ataxin 1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-45237
103408-280EA 564.74 USD
103408-280
Anti-Ataxin 1 Goat Polyclonal Antibody
Antibodies
The Ataxin 1 Antibody from Novus Biologicals is a goat polyclonal antibody to Ataxin 1. This antibody reacts with human, porcine, canine. The Ataxin 1 Antibody has been validated for the following applications: Peptide ELISA.

Peptide

Type: Primary
Antigen: Ataxin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal region
Host: Goat
Isotype: IgG
Reactivity: Human, Porcine, Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR