Anti-Ataxin 1 Mouse Monoclonal Antibody [clone: 4C5]

Supplier: Novus Biologicals
H00006310-M02
103309-806EA 798.46 USD
103309-806
Anti-Ataxin 1 Mouse Monoclonal Antibody [clone: 4C5]
Antibodies
The Ataxin 1 Antibody (4C5) from Novus Biologicals is a mouse monoclonal antibody to Ataxin 1. This antibody reacts with human. The Ataxin 1 Antibody (4C5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Ataxin 1
Clonality: Monoclonal
Clone: 4C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR