|
![]() |
ORDER
SPECIFICATIONS
Antigen Name | Astrovirus, serotype 1 |
Reactivity | Virus |
Conjugation | DyLight® 405 |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | ASV |
Host | Mouse |
Isotype | IgG3 |
Western Blot | Yes |
ELISA | Yes |
Immunogen | Astrovirus, serotype 1 |
Purification | Protein G purified |
Storage Buffer | 50mM Sodium Borate |
Storage Temperature | Store at 4C in the dark. |