Anti-Astrovirus, serotype 1 Mouse Monoclonal Antibody (Biotin) [clone: ASV]

Supplier: Novus Biologicals
NB100-73099B
103401-772EA 399 USD
103401-772
Anti-Astrovirus, serotype 1 Mouse Monoclonal Antibody (Biotin) [clone: ASV]
Antibodies
The Astrovirus, serotype 1 Antibody (ASV) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to Astrovirus, serotype 1. This antibody reacts with virus. The Astrovirus, serotype 1 Antibody (ASV) [Biotin] has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Astrovirus, serotype 1
Clonality: Monoclonal
Clone: ASV
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR