Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Astrovirus Mouse Monoclonal Antibody (DyLight 755) [clone: BGN/07/8630]

Supplier: Novus Biologicals
NB100-62361IR
103401-756
Anti-Astrovirus Mouse Monoclonal Antibody (DyLight 755) [clone: BGN/07/8630]
The Astrovirus Antibody (BGN / 07 / 8630) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to Astrovirus. This antibody reacts with virus. The Astrovirus Antibody (BGN / 07 / 8630) [DyLight 755] has been validated for the following applications: ELISA.

Type: Primary
Antigen: Astrovirus
Clonality: Monoclonal
Clone: BGN/07/8630
Conjugation: DyLight 755
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Virus

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty