Anti-Astrovirus Mouse Monoclonal Antibody (DyLight 350) [clone: BGN/07/8611]

NB100-62358UV
103403-682EA 423.08 USD
103403-682
Anti-Astrovirus Mouse Monoclonal Antibody (DyLight 350) [clone: BGN/07/8611]
The Astrovirus Antibody (BGN / 07 / 8611) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to Astrovirus. This antibody reacts with virus. The Astrovirus Antibody (BGN / 07 / 8611) [DyLight 350] has been validated for the following applications: ELISA.

Type: Primary
Antigen: Astrovirus
Clonality: Monoclonal
Clone: BGN/07/8611
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR