Anti-Asporin Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1514SS
103408-278EA 129 USD
103408-278
Anti-Asporin Goat Polyclonal Antibody
Antibodies
The Asporin Antibody from Novus Biologicals is a goat polyclonal antibody to Asporin. This antibody reacts with human, mouse, rat. The Asporin Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Peptide ELISA, Immunocytochemistry.

Type: Primary
Antigen: Asporin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR