Anti-Aspergillus Mouse Monoclonal Antibody (DyLight 488) [clone: 343/31]

NB100-64511G
103403-664EA 536.92 USD
103403-664
Anti-Aspergillus Mouse Monoclonal Antibody (DyLight 488) [clone: 343/31]
The Aspergillus Antibody (343 / 31) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to Aspergillus. This antibody reacts with fungi. The Aspergillus Antibody (343 / 31) [DyLight 488] has been validated for the following applications: ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Aspergillus
Clonality: Monoclonal
Clone: 343/31
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Fungi
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR