Anti-Aspartyl Aminopeptidase Mouse Monoclonal Antibody [clone: 2F9-3A7]

Supplier: Novus Biologicals
H00023549-M01
103341-492EA 798.46 USD
103341-492
Anti-Aspartyl Aminopeptidase Mouse Monoclonal Antibody [clone: 2F9-3A7]
Antibodies
The Aspartyl Aminopeptidase Antibody (2F9-3A7) from Novus Biologicals is a mouse monoclonal antibody to Aspartyl Aminopeptidase. This antibody reacts with human. The Aspartyl Aminopeptidase Antibody (2F9-3A7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Aspartyl Aminopeptidase
Clonality: Monoclonal
Clone: 2F9-3A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR