Anti-Aspartate beta hydroxylase Mouse Polyclonal Antibody

H00000444-B01
103322-242EA 583.08 USD
103322-242
Anti-Aspartate beta hydroxylase Mouse Polyclonal Antibody
The Aspartate beta hydroxylase Antibody from Novus Biologicals is a mouse polyclonal antibody to Aspartate beta hydroxylase. This antibody reacts with human. The Aspartate beta hydroxylase Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Aspartate beta hydroxylase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR