Anti-Aspartate beta hydroxylase Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00000444-D03P
103322-250EA 429 USD
103322-250
Anti-Aspartate beta hydroxylase Rabbit Polyclonal Antibody
Antibodies
The Aspartate beta hydroxylase Antibody from Novus Biologicals is a rabbit polyclonal antibody to Aspartate beta hydroxylase. This antibody reacts with human. The Aspartate beta hydroxylase Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Aspartate beta hydroxylase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR