Anti-Asparagine synthetase Mouse Monoclonal Antibody [clone: 2B3]

Supplier: Novus Biologicals
H00000440-M02
103322-234EA 798.46 USD
103322-234
Anti-Asparagine synthetase Mouse Monoclonal Antibody [clone: 2B3]
Antibodies
The Asparagine synthetase Antibody (2B3) from Novus Biologicals is a mouse monoclonal antibody to Asparagine synthetase. This antibody reacts with human. The Asparagine synthetase Antibody (2B3) has been validated for the following applications: ELISA, Immunoprecipitation.

Type: Primary
Antigen: Asparagine synthetase
Clonality: Monoclonal
Clone: 2B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR