Anti-Asialoglycoprotein Receptor 2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00000433-B01P
103322-208EA 767.69 USD
103322-208
Anti-Asialoglycoprotein Receptor 2 Mouse Polyclonal Antibody
Antibodies
The Asialoglycoprotein Receptor 2 Antibody from Novus Biologicals is a mouse polyclonal antibody to Asialoglycoprotein Receptor 2. This antibody reacts with human. The Asialoglycoprotein Receptor 2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Asialoglycoprotein Receptor 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR