Anti-Arginyl tRNA synthetase Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15448
102705-998EA 635.51 USD
102705-998
Anti-Arginyl tRNA synthetase Rabbit Polyclonal Antibody
Antibodies
The Arginyl tRNA synthetase Antibody from Novus Biologicals is a rabbit polyclonal antibody to Arginyl tRNA synthetase. This antibody reacts with human. The Arginyl tRNA synthetase Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Arginyl tRNA synthetase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR