Anti-Aquaporin-7 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00000364-B01P
103322-022EA 798.46 USD
103322-022
Anti-Aquaporin-7 Mouse Polyclonal Antibody
Antibodies
The Aquaporin-7 Antibody from Novus Biologicals is a mouse polyclonal antibody to Aquaporin-7. This antibody reacts with human. The Aquaporin-7 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Aquaporin-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR