Anti-Antizyme inhibitor 1 Mouse Monoclonal Antibody [clone: 1F11]

Supplier: Novus Biologicals
H00051582-M02
103344-316EA 798.46 USD
103344-316
Anti-Antizyme inhibitor 1 Mouse Monoclonal Antibody [clone: 1F11]
Antibodies
The Antizyme inhibitor 1 Antibody (1F11) from Novus Biologicals is a mouse monoclonal antibody to Antizyme inhibitor 1. This antibody reacts with human. The Antizyme inhibitor 1 Antibody (1F11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Antizyme inhibitor 1
Clonality: Monoclonal
Clone: 1F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR