Anti-Ankyrin repeat domain 39 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15402
102705-906EA 635.51 USD
102705-906
Anti-Ankyrin repeat domain 39 Rabbit Polyclonal Antibody
Antibodies
The Ankyrin repeat domain 39 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Ankyrin repeat domain 39. This antibody reacts with human. The Ankyrin repeat domain 39 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Ankyrin repeat domain 39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR